siRNA Details
Gene Symbol: XCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003175.3 | NP_003166.1 | 3 | 301 | ||
GenBank mRNA | BC069360.1 | - | 3 | 268 | 7262751 | |
BC070308.1 | - | 3 | 301 | 30413788 | ||
CD521967.1 | - | 3 | 304 |
Gene Symbol: XCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002995.2 | NP_002986.1 | 3 | 433 | ||
XM_011509865.2 | XP_011508167.1 | 4 | 2658 | |||
GenBank mRNA | BC069817.1 | - | 3 | 286 | 7262708 | |
BC070309.1 | - | 3 | 301 | 30416260 | ||
BF433809.1 | - | 3 | 433 | |||
D43768.1 | - | 3 | 288 | |||
U23772.1 | - | 3 | 282 | |||
X86474.1 | - | 3 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
