siRNA Details
Gene Symbol: LOC645166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027354.2 | 2 | 245 | |||
NR_027355.2 | 2 | 145 | ||||
NR_027356.2 | 2 | 145 | ||||
GenBank mRNA | AK311729.1 | - | 2 | 245 | ||
AW954772.1 | - | 2 | 134 | |||
BG716766.1 | - | 2 | 137 | |||
BQ438821.1 | - | 2 | 146 | |||
CR746216.1 | - | 2 | 172 | |||
DB496177.1 | - | 2 | 132 |
Gene Symbol: LOC654342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030764.1 | - | 2 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
