siRNA Details
Gene Symbol: LSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013253.1 | NP_001013271.1 | 8 | 1073 | ||
NM_001013254.1 | NP_001013272.1 | 8 | 1303 | |||
NM_001013255.1 | NP_001013273.1 | 8 | 809 | |||
NM_001242932.1 | NP_001229861.1 | 9 | 1304 | |||
NM_001289005.1 | NP_001275934.1 | 8 | 1169 | |||
NM_002339.2 | NP_002330.1 | 8 | 987 | |||
GenBank mRNA | AK056576.1 | - | 8 | 1928 | ||
AK092071.1 | - | 9 | 1304 | |||
AK093859.1 | - | 7 | 1310 | |||
AK222733.1 | - | 8 | 883 | |||
AK290279.1 | - | 8 | 825 | |||
AK292033.1 | - | 8 | 1171 | |||
BC001785.1 | - | 8 | 879 | 3542307 | ||
CR541728.1 | - | 8 | 812 | |||
CR541754.1 | - | 8 | 812 | |||
S67783.1 | - | 8 | 809 | |||
X55188.1 | - | 8 | 859 |
Gene Symbol: LOC107985911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005465.1 | XP_016860954.1 | 2 | 507 |
Gene Symbol: LOC645166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW954772.1 | - | 2 | 157 | ||
BE814857.1 | - | 2 | 160 | |||
CR746216.1 | - | 2 | 195 | |||
DA808118.1 | - | 2 | 170 | |||
DB496177.1 | - | 2 | 155 |
Gene Symbol: LOC654342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030764.1 | - | 2 | 160 |
Gene Symbol: LSP1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033961.1 | 1 | 41 | |||
GenBank mRNA | BC121073.2 | - | 1 | 45 | 40119616 | |
BC121074.2 | - | 1 | 45 | 40119620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
