siRNA Details
Gene Symbol: CCZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015622.5 | NP_056437.4 | 1, 2 | 212 | ||
GenBank mRNA | AF151801.1 | - | 1, 2 | 256 | ||
BC067810.1 | - | 1, 2 | 222 | |||
BC132749.1 | - | 1, 2 | 197 | 40146771 | ||
BC132751.1 | - | 1, 2 | 197 | 40146773 | ||
BI546694.1 | - | 1, 2 | 204 |
Gene Symbol: CCZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198097.3 | NP_932765.1 | 1, 2 | 220 | ||
GenBank mRNA | AK000993.1 | - | 1, 2 | 141 | ||
AK292326.1 | - | 1, 2 | 188 | |||
AK296026.1 | - | 1, 2 | 120 | |||
BC010130.2 | - | 1, 2 | 155 | 3953739 | ||
BP341272.1 | - | 1, 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
