siRNA Details
Gene Symbol: LOC105374313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136187.1 | 3 | 554 |
Gene Symbol: RPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000994.3 | NP_000985.1 | 3, 4 | 371 | ||
NM_001007073.1 | NP_001007074.1 | 3, 4 | 470 | |||
NM_001007074.1 | NP_001007075.1 | 2, 3 | 490 | |||
GenBank mRNA | AK311909.1 | - | 3, 4 | 328 | ||
BC011514.1 | - | 1 | 297 | 2900089 | ||
BC070209.1 | - | 1 | 303 | 6738371 | ||
BQ068496.1 | - | 408 | ||||
CF124158.1 | - | 3, 4 | 408 | |||
X03342.1 | - | 3, 4 | 311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
