siRNA Details
Gene Symbol: RPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000994.3 | NP_000985.1 | 2 | 143 | ||
NM_001007073.1 | NP_001007074.1 | 2 | 242 | |||
NM_001007074.1 | NP_001007075.1 | 1 | 262 | |||
GenBank mRNA | AK311909.1 | - | 2 | 100 | ||
BC011514.1 | - | 1 | 69 | 2900089 | ||
BC070209.1 | - | 1 | 75 | 6738371 | ||
BF960792.1 | - | 1 | 273 | |||
BQ068496.1 | - | 180 | ||||
CF124158.1 | - | 2 | 180 | |||
X03342.1 | - | 2 | 83 |
Gene Symbol: MTHFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010702.1 | XP_016866191.1 | 28 | 3130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
