siRNA Details
Gene Symbol: TTTY7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001534.2 | 7 | 1153 | |||
GenBank mRNA | AF278762.1 | - | 3 | 495 |
Gene Symbol: TTTY7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003592.1 | 6 | 897 | |||
GenBank mRNA | BX089948.1 | - | 3 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
