siRNA Details
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 6, 7 | 695 | ||
NM_001145356.1 | NP_001138828.1 | 5, 6 | 597 | |||
NM_018491.3 | NP_060961.3 | 5, 6 | 597 | |||
XM_005251514.1 | XP_005251571.1 | 5, 6 | 597 | |||
XM_005251515.1 | XP_005251572.1 | 5, 6 | 597 | |||
XM_005251517.4 | XP_005251574.1 | 5, 6 | 597 | |||
XM_011517961.2 | XP_011516263.1 | 6, 7 | 689 | |||
XM_011517962.2 | XP_011516264.1 | 5, 6 | 597 | |||
XM_011517963.1 | XP_011516265.1 | 2, 3 | 135 | |||
XM_011517964.2 | XP_011516266.1 | 5, 6 | 597 | |||
XM_017014925.1 | XP_016870414.1 | 5, 6 | 597 | |||
GenBank mRNA | AF212253.1 | - | 5, 6 | 565 | ||
AK290648.1 | - | 6, 7 | 667 | |||
AK297292.1 | - | 6, 7 | 636 | |||
AK315855.1 | - | 5, 6 | 521 | |||
AY295766.1 | - | 5, 6 | 455 | |||
AY343911.1 | - | 5, 6 | 554 | |||
BC005996.1 | - | 5, 6 | 597 | 4069589 | ||
BC009573.1 | - | 6, 7 | 648 | 4133536 | ||
BC013432.1 | - | 5, 6 | 568 | 4133722 | ||
BC132759.1 | - | 5, 6 | 531 | 40146781 |

More Information
Other Species Targeted

Gene Ontology Categories:
