siRNA Details
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 9 | 804 | ||
NM_001145356.1 | NP_001138828.1 | 8 | 706 | |||
NM_018491.3 | NP_060961.3 | 8 | 706 | |||
XM_005251513.4 | XP_005251570.1 | 7 | 654 | |||
XM_005251517.4 | XP_005251574.1 | 8 | 706 | |||
XM_011517961.2 | XP_011516263.1 | 9 | 798 | |||
XM_011517962.2 | XP_011516264.1 | 8 | 706 | |||
XM_011517963.1 | XP_011516265.1 | 5 | 244 | |||
XM_017014925.1 | XP_016870414.1 | 8 | 706 | |||
GenBank mRNA | AF212253.1 | - | 8 | 674 | ||
AF257330.2 | - | 8 | 664 | |||
AK290648.1 | - | 9 | 776 | |||
AK297292.1 | - | 9 | 745 | |||
AK315855.1 | - | 8 | 630 | |||
AY343911.1 | - | 8 | 663 | |||
BC005996.1 | - | 8 | 706 | 4069589 | ||
BC009573.1 | - | 9 | 757 | 4133536 | ||
BC013432.1 | - | 8 | 677 | 4133722 | ||
BC132759.1 | - | 8 | 640 | 40146781 |
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 8 | 778 | ||
XM_005263600.2 | XP_005263657.1 | 8 | 778 | |||
XM_005263603.2 | XP_005263660.1 | 9 | 812 | |||
XM_005263605.3 | XP_005263662.1 | 9 | 813 | |||
XM_011510667.1 | XP_011508969.1 | 8 | 778 | |||
XM_011510668.2 | XP_011508970.1 | 5 | 338 | |||
XM_011510669.2 | XP_011508971.1 | 9 | 822 | |||
XM_017003407.1 | XP_016858896.1 | 8 | 778 | |||
XM_017003408.1 | XP_016858897.1 | 10 | 942 | |||
GenBank mRNA | AF452722.1 | - | 8 | 634 | ||
AK225251.1 | - | 9 | 792 | |||
AK290072.1 | - | 8 | 778 | |||
AK303653.1 | - | 9 | 817 | |||
AY343910.1 | - | 8 | 663 | |||
BC120991.1 | - | 8 | 649 | 40116409 | ||
BC120992.1 | - | 8 | 649 | 40116411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
