siRNA Details
Gene Symbol: CBWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145355.1 | NP_001138827.1 | 9 | 828 | ||
NM_001145356.1 | NP_001138828.1 | 8 | 730 | |||
NM_018491.3 | NP_060961.3 | 8 | 730 | |||
XM_005251513.4 | XP_005251570.1 | 7 | 678 | |||
XM_005251517.4 | XP_005251574.1 | 8 | 730 | |||
XM_011517961.2 | XP_011516263.1 | 9 | 822 | |||
XM_011517962.2 | XP_011516264.1 | 8 | 730 | |||
XM_011517963.1 | XP_011516265.1 | 5 | 268 | |||
XM_017014925.1 | XP_016870414.1 | 8 | 730 | |||
GenBank mRNA | AF212253.1 | - | 8 | 698 | ||
AF257330.2 | - | 8 | 688 | |||
AK290648.1 | - | 9 | 800 | |||
AK297292.1 | - | 9 | 769 | |||
AK315855.1 | - | 8 | 654 | |||
AY343911.1 | - | 8 | 687 | |||
BC005996.1 | - | 8 | 730 | 4069589 | ||
BC009573.1 | - | 9 | 781 | 4133536 | ||
BC013432.1 | - | 8 | 701 | 4133722 | ||
BC132759.1 | - | 8 | 664 | 40146781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
