siRNA Details
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012423.3 | NP_036555.1 | 3, 4 | 228 | ||
NR_073024.1 | 3, 4 | 218 | ||||
GenBank mRNA | AK130605.1 | - | 3, 4 | 175 | ||
AK222747.1 | - | 3, 4 | 154 | |||
AK291120.1 | - | 3, 4 | 174 | |||
AK297859.1 | - | 3, 4 | 174 | |||
BC000514.2 | - | 3, 4 | 157 | 2822802 | ||
BC001675.2 | - | 1 | 155 | 2961667 | ||
BC001836.2 | - | 1 | 155 | 2961667 | ||
BC004900.2 | - | 2, 3 | 148 | |||
BC013230.1 | - | 1 | 150 | |||
BC032107.2 | - | 3, 4 | 163 | |||
BC062537.1 | - | 2, 3 | 154 | 4908022 | ||
BC065236.1 | - | 1 | 162 | 6161599 | ||
BC070223.1 | - | 3, 4 | 174 | 6736442 | ||
BC071929.1 | - | 2, 3 | 158 | 5808487 | ||
BC105605.1 | - | 1 | 168 | 6726477 | ||
BG536330.1 | - | 3, 4 | 175 | |||
X56932.1 | - | 3, 4 | 169 |
Gene Symbol: RPL13AP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003932.2 | 1 | 174 |
Gene Symbol: RPL13AP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026712.1 | 1 | 174 |
Gene Symbol: RPL13AP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026715.1 | 1 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
