siRNA Details
Gene Symbol: RPL13AP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026712.1 | 1 | 209 |
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270491.1 | NP_001257420.1 | 3 | 187 | ||
NM_012423.3 | NP_036555.1 | 4 | 263 | |||
NR_073024.1 | 4 | 253 | ||||
GenBank mRNA | AB082924.1 | - | 1 | 42 | ||
AK056837.1 | - | 2 | 695 | |||
AK130605.1 | - | 4 | 210 | |||
AK222747.1 | - | 4 | 189 | |||
AK291120.1 | - | 4 | 209 | |||
AK297859.1 | - | 4 | 209 | |||
BC000514.2 | - | 4 | 192 | 2822802 | ||
BC001675.2 | - | 1 | 190 | 2961667 | ||
BC001836.2 | - | 1 | 190 | 2961667 | ||
BC004900.2 | - | 3 | 183 | |||
BC013230.1 | - | 1 | 185 | |||
BC032107.2 | - | 4 | 198 | |||
BC062537.1 | - | 3 | 189 | 4908022 | ||
BC065236.1 | - | 1 | 197 | 6161599 | ||
BC070223.1 | - | 4 | 209 | 6736442 | ||
BC071929.1 | - | 3 | 193 | 5808487 | ||
BC105605.1 | - | 1 | 203 | 6726477 | ||
BG536330.1 | - | 4 | 210 | |||
BQ069994.1 | - | 2 | 114 | |||
X56932.1 | - | 4 | 204 |
Gene Symbol: RPL13AP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004844.1 | 1 | 119 | |||
GenBank mRNA | BC067891.1 | - | 1 | 119 |
Gene Symbol: RPL13AP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026715.1 | 1 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
