siRNA Details
Gene Symbol: FRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005217.1 | NP_001005217.1 | 1 | 141 | ||
NM_001286820.1 | NP_001273749.1 | 1 | 141 | |||
GenBank mRNA | AY714545.1 | - | 1 | 141 | ||
BC144572.1 | - | 1 | 126 | 9053101 |
Gene Symbol: FRG2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080998.1 | NP_001074467.1 | 1 | 141 | ||
GenBank mRNA | AY744466.1 | - | 1 | 141 |
Gene Symbol: FRG2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124759.1 | NP_001118231.1 | 1 | 132 | ||
XM_011533254.2 | XP_011531556.1 | 2 | 965 | |||
XM_011533256.2 | XP_011531558.1 | 2 | 965 |
Gene Symbol: FRG2DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026980.2 | 1, 2 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
