siRNA Details
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300795.1 | NP_001287724.1 | 7 | 503 | ||
NM_003340.6 | NP_003331.1 | 7 | 810 | |||
NM_181886.3 | NP_871615.1 | 7 | 1018 | |||
NM_181887.2 | NP_871616.1 | 7 | 788 | |||
NM_181888.3 | NP_871617.1 | 7 | 667 | |||
NM_181889.2 | NP_871618.1 | 7 | 690 | |||
NM_181890.2 | NP_871619.1 | 7 | 632 | |||
NM_181891.2 | NP_871620.1 | 7 | 975 | |||
NM_181892.3 | NP_871621.1 | 6 | 1000 | |||
NM_181893.2 | NP_871622.1 | 6 | 480 | |||
XM_017008584.1 | XP_016864073.1 | 6 | 842 | |||
GenBank mRNA | AK095822.1 | - | 6 | 915 | ||
AK127304.1 | - | 5 | 3423 | |||
AK315775.1 | - | 7 | 576 | |||
AL833414.1 | - | 6 | 1593 | |||
AU131013.1 | - | 7 | 578 | |||
BC003395.1 | - | 7 | 490 | 3447673 | ||
BC037894.2 | - | 7 | 787 | 5275356 | ||
BC066917.1 | - | 7 | 521 | 4792462 | ||
BG613555.1 | - | 7 | 616 | |||
BM541512.1 | - | 7 | 772 | |||
BQ428985.1 | - | 7 | 485 | |||
BU567649.1 | - | 7 | 660 | |||
BU570661.1 | - | 6 | 435 | |||
CR456916.1 | - | 6 | 325 | |||
DA335600.1 | - | 7 | 568 | |||
DA965437.1 | - | 6 | 486 | |||
DB234695.1 | - | 6 | 424 | |||
U39318.1 | - | 6 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
