siRNA Details
Gene Symbol: C2orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182500.2 | NP_872306.1 | 3 | 665 | ||
XM_006711857.2 | XP_006711920.1 | 3 | 589 | |||
XM_006711858.2 | XP_006711921.1 | 3 | 589 | |||
XM_006711859.3 | XP_006711922.1 | 3 | 589 | |||
GenBank mRNA | AK057872.1 | - | 3 | 671 | ||
AK292828.1 | - | 3 | 590 | |||
BC120930.2 | - | 3 | 632 | 40114857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
