siRNA Details
Gene Symbol: FAM86C2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024249.1 | 2 | 134 |
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289029.1 | NP_001275958.1 | 2 | 192 | ||
NM_201400.3 | NP_958802.1 | 2 | 192 | |||
NM_201598.3 | NP_963892.1 | 2 | 192 | |||
XM_005255157.4 | XP_005255214.1 | 2 | 165 | |||
XM_005255158.4 | XP_005255215.1 | 3 | 238 | |||
XM_011522404.2 | XP_011520706.1 | 3 | 238 | |||
GenBank mRNA | AL522535.3 | - | 2 | 114 | ||
AY037162.1 | - | 2 | 160 | |||
BC010084.2 | - | 2 | 112 | 2822323 | ||
BG024981.1 | - | 2 | 160 | |||
BQ921410.1 | - | 2 | 175 | |||
CA495371.1 | - | 2 | 113 | |||
HY017433.1 | - | 2 | 192 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 2 | 137 | ||
NM_018172.2 | NP_060642.2 | 2 | 137 | |||
NM_152563.2 | NP_689776.1 | 2 | 137 | |||
GenBank mRNA | AK001523.1 | - | 2 | 111 | ||
AK130709.1 | - | 2 | 134 | |||
AK293221.1 | - | 2 | 111 | |||
AK311428.1 | - | 2 | 111 | |||
AK311518.1 | - | 2 | 134 | |||
AK311675.1 | - | 2 | 138 | |||
AK315454.1 | - | 2 | 111 | |||
BC032519.1 | - | 2 | 137 | 5494088 | ||
BQ921333.1 | - | 2 | 107 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 2 | 213 | |||
GenBank mRNA | BC043342.2 | - | 2 | 39 | ||
BC068108.1 | - | 2 | 146 | |||
BC094682.1 | - | 2 | 260 | |||
DA753838.1 | - | 2 | 213 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 2 | 120 | |||
NR_130741.1 | 2 | 120 | ||||
NR_130742.1 | 2 | 120 | ||||
GenBank mRNA | AA421090.1 | - | 2 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
