siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 3 | 238 | ||
NR_003494.1 | 3 | 238 | ||||
XM_006716256.3 | XP_006716319.1 | 3 | 231 | |||
XM_006716262.1 | XP_006716325.1 | 3 | 114 | |||
XM_011543842.2 | XP_011542144.1 | 3 | 188 | |||
XM_017013916.1 | XP_016869405.1 | 3 | 237 | |||
GenBank mRNA | AK098138.1 | - | 3 | 205 | ||
AK308439.1 | - | 3 | 217 | |||
BC062615.1 | - | 3 | 283 | |||
BC085016.1 | - | 4 | 311 | |||
BC098427.1 | - | 3 | 288 | |||
BE729618.1 | - | 3 | 180 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 3 | 193 | ||
XM_005273603.4 | XP_005273660.1 | 3 | 268 | |||
XM_005273607.2 | XP_005273664.1 | 3 | 250 | |||
XM_011544618.2 | XP_011542920.1 | 3 | 238 | |||
XM_011544619.2 | XP_011542921.1 | 2 | 205 | |||
XM_017013740.1 | XP_016869229.1 | 3 | 188 | |||
XM_017013741.1 | XP_016869230.1 | 2 | 125 | |||
GenBank mRNA | AK291001.1 | - | 3 | 250 |
Gene Symbol: FAM86B3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024361.1 | 3 | 228 | |||
NR_024362.1 | 3 | 325 | ||||
NR_024363.1 | 3 | 325 | ||||
GenBank mRNA | AK304567.1 | - | 3 | 297 | ||
AK307256.1 | - | 3 | 295 | |||
BC035792.1 | - | 3 | 203 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 3 | 219 | ||
GenBank mRNA | AK311428.1 | - | 3 | 196 | ||
AK311518.1 | - | 3 | 219 | |||
AK311675.1 | - | 3 | 223 | |||
BQ921333.1 | - | 3 | 189 | |||
DC312827.1 | - | 3 | 222 |
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024241.1 | 3 | 297 | |||
GenBank mRNA | BC043342.2 | - | 3 | 123 | ||
BC068108.1 | - | 3 | 230 | |||
BC094682.1 | - | 3 | 344 | |||
DA753838.1 | - | 3 | 297 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130742.1 | 3 | 202 | |||
GenBank mRNA | AA421090.1 | - | 3 | 165 | ||
CN271531.1 | - | 4 | 242 | |||
HY065247.1 | - | 3 | 202 |
Gene Symbol: FAM86FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024254.1 | 2 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
