siRNA Details
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098138.1 | - | 4 | 270 | ||
AK308439.1 | - | 5 | 384 | |||
AL529666.3 | - | 3 | 240 | |||
BC062615.1 | - | 4 | 348 | |||
BC085016.1 | - | 5 | 376 | |||
BE729618.1 | - | 4 | 245 |
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK291001.1 | - | 4 | 315 | ||
AK309984.1 | - | 5 | 413 |
Gene Symbol: FAM86EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130740.1 | 3 | 186 | |||
NR_130741.1 | 3 | 186 | ||||
GenBank mRNA | AA421090.1 | - | 5 | 375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
