siRNA Details
Gene Symbol: MZT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025029.3 | NP_079305.2 | 2 | 546 | ||
XM_011511886.2 | XP_011510188.1 | 2 | 290 | |||
XM_011511887.2 | XP_011510189.1 | 2 | 257 | |||
XM_011511888.2 | XP_011510190.1 | 2 | 591 | |||
XM_011511889.2 | XP_011510191.1 | 2 | 290 | |||
XM_011511890.2 | XP_011510192.1 | 2 | 290 | |||
GenBank mRNA | BC014255.2 | - | 2 | 188 | ||
BC066296.1 | - | 2 | 278 | 5284352 | ||
BG036944.1 | - | 2 | 324 |
Gene Symbol: MZT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085365.1 | NP_001078834.1 | 2 | 306 | ||
XM_005263741.4 | XP_005263798.1 | 2 | 534 | |||
XM_005263742.2 | XP_005263799.1 | 2 | 534 | |||
XM_017004744.1 | XP_016860233.1 | 2 | 534 | |||
GenBank mRNA | AK090531.1 | - | 1 | 90 | ||
BC018206.1 | - | 2 | 210 | 3862861 | ||
BC063024.1 | - | 2 | 189 | |||
BC104650.1 | - | 1 | 87 | |||
BI599271.1 | - | 2 | 540 | |||
BM554057.1 | - | 2 | 368 |
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ224347.1 | - | 214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
