siRNA Details
Gene Symbol: MZT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025029.3 | NP_079305.2 | 3 | 887 | ||
XM_005263795.4 | XP_005263852.1 | 2 | 792 | |||
XM_011511886.2 | XP_011510188.1 | 4 | 811 | |||
XM_011511887.2 | XP_011510189.1 | 4 | 778 | |||
XM_011511888.2 | XP_011510190.1 | 4 | 1112 | |||
GenBank mRNA | BC014255.2 | - | 3 | 529 | ||
BC066296.1 | - | 3 | 619 | 5284352 |
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ224347.1 | - | 555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
