siRNA Details
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321170.1 | NP_001308099.1 | 9 | 736 | ||
NM_138501.5 | NP_612510.1 | 8 | 662 | |||
NR_038103.1 | 8 | 694 | ||||
NR_038104.1 | 8 | 727 | ||||
XM_006722945.1 | XP_006723008.1 | 9 | 780 | |||
XM_011528442.2 | XP_011526744.1 | 8 | 610 | |||
GenBank mRNA | AF038958.1 | - | 8 | 608 | ||
AF038959.1 | - | 3 | 91 | |||
AF222742.1 | - | 8 | 583 | |||
AK001416.1 | - | 8 | 626 | |||
AK094087.1 | - | 9 | 703 | |||
AK299134.1 | - | 9 | 700 | |||
AK315412.1 | - | 8 | 626 | |||
BC000174.2 | - | 7 | 522 | |||
BC002425.1 | - | 8 | 594 | 3163592 | ||
BC005952.1 | - | 8 | 605 | 4249841 | ||
BC007801.2 | - | 8 | 612 | 4128702 | ||
BC013881.2 | - | 8 | 610 | 3928698 | ||
BG705196.1 | - | 8 | 665 | |||
BT007179.1 | - | 7 | 532 | |||
CR456892.1 | - | 7 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
