siRNA Details
Gene Symbol: GOLGA8IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024074.1 | 12 | 1586 | |||
GenBank mRNA | AK093104.1 | - | 12 | 1586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
