siRNA Details
Gene Symbol: GOLGA8M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282468.1 | NP_001269397.1 | 7, 8 | 576 | ||
XM_005268284.4 | XP_005268341.1 | 7, 8 | 1375 | |||
XM_005268285.4 | XP_005268342.1 | 7, 8 | 1375 | |||
XM_006720654.3 | XP_006720717.1 | 7, 8 | 519 | |||
XM_011521951.2 | XP_011520253.1 | 8, 9 | 944 | |||
XM_011521952.2 | XP_011520254.1 | 7, 8 | 1375 | |||
XM_011521953.2 | XP_011520255.1 | 7, 8 | 1375 | |||
XM_011521954.2 | XP_011520256.1 | 7, 8 | 509 |
Gene Symbol: GOLGA8H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282490.1 | NP_001269419.1 | 7, 8 | 574 |
Gene Symbol: GOLGA8IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024074.1 | 6, 7 | 777 | |||
GenBank mRNA | AK093104.1 | - | 6, 7 | 777 |
Gene Symbol: GOLGA8J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282472.1 | NP_001269401.1 | 7, 8 | 576 | ||
XM_006720651.3 | XP_006720714.1 | 7, 8 | 596 | |||
XM_011521948.2 | XP_011520250.1 | 7, 8 | 904 | |||
XM_011521950.2 | XP_011520252.1 | 7, 8 | 902 | |||
GenBank mRNA | DB054802.1 | - | 7, 8 | 465 |
Gene Symbol: GOLGA8K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282493.1 | NP_001269422.1 | 7, 8 | 576 |
Gene Symbol: GOLGA8T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033933.1 | 7, 8 | 596 | |||
GenBank mRNA | AK310526.1 | - | 7, 8 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
