siRNA Details
Gene Symbol: DISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032890.3 | NP_116279.2 | 10 | 1671 | ||
XM_005273335.2 | XP_005273392.1 | 8 | 1512 | |||
XM_006711592.2 | XP_006711655.1 | 9 | 2459 | |||
XM_006711594.3 | XP_006711657.1 | 8 | 1167 | |||
XM_011510072.2 | XP_011508374.1 | 9 | 1511 | |||
XM_011510073.2 | XP_011508375.1 | 9 | 1989 | |||
XM_011510074.2 | XP_011508376.1 | 9 | 1530 | |||
XM_011510075.2 | XP_011508377.1 | 9 | 2017 | |||
XM_011510077.2 | XP_011508379.1 | 8 | 1453 | |||
XM_017002611.1 | XP_016858100.1 | 8 | 1927 | |||
XM_017002612.1 | XP_016858101.1 | 9 | 1336 | |||
XM_017002613.1 | XP_016858102.1 | 7 | 1296 | |||
GenBank mRNA | AK056569.1 | - | 3 | 475 | ||
AK098669.1 | - | 8 | 1405 | |||
AK125728.1 | - | 8 | 1509 | |||
BC007734.2 | - | 8 | 1492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
