siRNA Details
Gene Symbol: LOC107987001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015381.1 | XP_016870870.1 | 1 | 685 | ||
XM_017015382.1 | XP_016870871.1 | 1 | 685 | |||
XM_017015383.1 | XP_016870872.1 | 1 | 685 |
Gene Symbol: FAM27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027422.1 | 1 | 253 | |||
GenBank mRNA | AK131029.1 | - | 1 | 136 |
Gene Symbol: FAM27C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027421.1 | 1 | 253 | |||
GenBank mRNA | BC002886.1 | - | 1 | 184 |
Gene Symbol: LOC102723847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC033389.1 | - | 1 | 133 |
Gene Symbol: MGC39584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038377.1 | 1 | 136 | |||
GenBank mRNA | BC029568.1 | - | 1 | 136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
