siRNA Details
Gene Symbol: UNC93B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030930.3 | NP_112192.2 | 10 | 1465 | ||
XM_011545290.1 | XP_011543592.1 | 8 | 1075 | |||
XM_011545291.2 | XP_011543593.1 | 9 | 1357 | |||
GenBank mRNA | AJ271326.1 | - | 10 | 1424 | ||
AJ422142.1 | - | 10 | 1433 | |||
AY007125.1 | - | 9 | 1252 | |||
BC025669.1 | - | 10 | 1411 | |||
BC033623.1 | - | 10 | 1463 | |||
BC092472.1 | - | 10 | 1433 | 30348121 | ||
BC101568.1 | - | 10 | 1409 | 8069074 | ||
BC105104.1 | - | 10 | 1409 | 8144107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
