siRNA Details
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 7 | 797 | |||
GenBank mRNA | AK092583.1 | - | 6 | 672 | ||
AK294377.1 | - | 5 | 930 | |||
AK296182.1 | - | 8 | 933 | |||
AK309926.1 | - | 7 | 784 | |||
AK310121.1 | - | 7 | 671 | |||
AK310139.1 | - | 8 | 825 | |||
AK316487.1 | - | 6 | 986 | |||
BC063470.1 | - | 4 | 710 | |||
BC094698.1 | - | 4 | 1572 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 7 | 898 | ||
XM_011517658.2 | XP_011515960.1 | 7 | 1015 | |||
XM_011517659.2 | XP_011515961.1 | 7 | 797 | |||
XM_011517660.2 | XP_011515962.1 | 7 | 794 | |||
XM_011517662.2 | XP_011515964.1 | 7 | 2301 | |||
XM_011517663.2 | XP_011515965.1 | 6 | 2150 | |||
XM_011517664.2 | XP_011515966.1 | 6 | 2147 | |||
XM_011517665.2 | XP_011515967.1 | 6 | 696 | |||
XM_011517666.2 | XP_011515968.1 | 6 | 695 | |||
XM_011517671.2 | XP_011515973.2 | 6 | 879 | |||
XM_017014169.1 | XP_016869658.1 | 7 | 914 | |||
XM_017014170.1 | XP_016869659.1 | 7 | 977 | |||
XM_017014171.1 | XP_016869660.1 | 5 | 589 | |||
XM_017014172.1 | XP_016869661.1 | 5 | 2167 | |||
XM_017014173.1 | XP_016869662.1 | 6 | 879 | |||
GenBank mRNA | AK024481.1 | - | 1 | 1752 | ||
AK289708.1 | - | 7 | 901 | |||
AK309568.1 | - | 5 | 869 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 7 | 1023 | |||
GenBank mRNA | AK292759.1 | - | 6 | 626 | ||
AY217347.1 | - | 6 | 1455 | |||
AY341936.1 | - | 2 | 152 | |||
AY341937.1 | - | 2 | 152 | |||
BC082240.1 | - | 4 | 1718 | |||
BC106020.1 | - | 4 | 447 | |||
BX377099.2 | - | 6 | 650 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 7 | 1572 | |||
GenBank mRNA | AK056232.1 | - | 7 | 1417 | ||
AK095183.1 | - | 6 | 1018 | |||
AK291582.1 | - | 6 | 1033 | |||
BC048328.1 | - | 6 | 772 | |||
BC063555.1 | - | 6 | 1211 | |||
BC110996.1 | - | 7 | 1274 | |||
BQ722707.1 | - | 6 | 782 | |||
BQ935819.1 | - | 2 | 188 | |||
DA065132.1 | - | 1 | 415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
