siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 11 | 1812 | ||
XM_011517658.2 | XP_011515960.1 | 11 | 1929 | |||
XM_011517659.2 | XP_011515961.1 | 11 | 1711 | |||
XM_011517660.2 | XP_011515962.1 | 11 | 1708 | |||
XM_011517662.2 | XP_011515964.1 | 11 | 3215 | |||
XM_011517663.2 | XP_011515965.1 | 10 | 3064 | |||
XM_011517664.2 | XP_011515966.1 | 10 | 3061 | |||
XM_011517665.2 | XP_011515967.1 | 10 | 1610 | |||
XM_011517666.2 | XP_011515968.1 | 10 | 1609 | |||
XM_017014169.1 | XP_016869658.1 | 11 | 1828 | |||
XM_017014170.1 | XP_016869659.1 | 11 | 1891 | |||
XM_017014171.1 | XP_016869660.1 | 9 | 1503 | |||
XM_017014172.1 | XP_016869661.1 | 9 | 3081 | |||
GenBank mRNA | AK024481.1 | - | 2 | 4220 | ||
BC094860.1 | - | 3 | 853 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 11 | 1814 | |||
GenBank mRNA | BC063682.1 | - | 7 | 1160 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 11 | 1940 | |||
GenBank mRNA | AL137714.1 | - | 8 | 3480 | ||
AY217347.1 | - | 10 | 2367 | |||
BC015400.1 | - | 3 | 632 | |||
BC073913.1 | - | 6 | 1329 | |||
BC082240.1 | - | 7 | 2775 | |||
BC093083.1 | - | 6 | 1342 | |||
BC106020.1 | - | 8 | 1377 | |||
BX537637.1 | - | 11 | 1579 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 11 | 2493 | |||
GenBank mRNA | AK056232.1 | - | 11 | 2338 | ||
AK057951.1 | - | 13 | 1938 | |||
AK095183.1 | - | 10 | 1934 | |||
AK097269.1 | - | 3 | 1719 | |||
AK290087.1 | - | 9 | 1554 | |||
BC048328.1 | - | 10 | 1702 | |||
BC063555.1 | - | 10 | 2128 | |||
BC110991.1 | - | 6 | 1102 | |||
BC110996.1 | - | 10 | 1979 | |||
BQ937971.1 | - | 1 | 334 |
Gene Symbol: WASH6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029274.1 | - | 2 | 1622 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 11 | 1708 | |||
GenBank mRNA | AK092583.1 | - | 7 | 3145 | ||
BC063470.1 | - | 7 | 1558 | |||
BC094698.1 | - | 8 | 2489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
