siRNA Details
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 10 | 1372 | |||
GenBank mRNA | AW274219.1 | - | 1 | 63 | ||
AW881719.1 | - | 2 | 549 | |||
BC063682.1 | - | 6 | 718 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 10 | 1372 | ||
XM_011517658.2 | XP_011515960.1 | 10 | 1489 | |||
XM_011517659.2 | XP_011515961.1 | 10 | 1271 | |||
XM_011517660.2 | XP_011515962.1 | 10 | 1268 | |||
XM_011517662.2 | XP_011515964.1 | 10 | 2775 | |||
XM_011517663.2 | XP_011515965.1 | 9 | 2624 | |||
XM_011517664.2 | XP_011515966.1 | 9 | 2621 | |||
XM_011517665.2 | XP_011515967.1 | 9 | 1170 | |||
XM_011517666.2 | XP_011515968.1 | 9 | 1169 | |||
XM_017014169.1 | XP_016869658.1 | 10 | 1388 | |||
XM_017014170.1 | XP_016869659.1 | 10 | 1451 | |||
XM_017014171.1 | XP_016869660.1 | 8 | 1063 | |||
XM_017014172.1 | XP_016869661.1 | 8 | 2641 | |||
GenBank mRNA | AK024481.1 | - | 2 | 3638 | ||
BC094860.1 | - | 2 | 413 | |||
CN404407.1 | - | 3 | 269 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 10 | 1493 | |||
GenBank mRNA | AK292759.1 | - | 9 | 1100 | ||
AL137714.1 | - | 7 | 3040 | |||
AY341936.1 | - | 5 | 626 | |||
AY341937.1 | - | 5 | 626 | |||
BC073913.1 | - | 5 | 889 | |||
BC082240.1 | - | 7 | 2188 | |||
BC093083.1 | - | 5 | 902 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 10 | 2046 | |||
GenBank mRNA | AK056232.1 | - | 10 | 1891 | ||
AK057951.1 | - | 12 | 1496 | |||
AK095183.1 | - | 9 | 1492 | |||
AK097269.1 | - | 2 | 1272 | |||
AK290087.1 | - | 8 | 1112 | |||
AK291582.1 | - | 9 | 1507 | |||
AW604905.1 | - | 2 | 138 | |||
BC063555.1 | - | 9 | 1681 | |||
BC110991.1 | - | 5 | 655 | |||
BQ935819.1 | - | 5 | 658 |
Gene Symbol: WASH6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029274.1 | - | 2 | 1040 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 10 | 1266 | |||
GenBank mRNA | AK092583.1 | - | 7 | 2558 | ||
BC094698.1 | - | 7 | 2042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
