siRNA Details
Gene Symbol: TRIM74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317815.1 | NP_001304744.1 | 4 | 848 | ||
NM_198853.2 | NP_942150.1 | 4 | 909 | |||
XM_011516187.2 | XP_011514489.1 | 4 | 817 | |||
XM_011516189.2 | XP_011514491.1 | 4 | 1133 | |||
XM_011516190.2 | XP_011514492.1 | 5 | 628 | |||
GenBank mRNA | AF498999.1 | - | 4 | 909 | ||
BC033871.1 | - | 4 | 848 | 5201777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
