siRNA Details
Gene Symbol: SSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009955.3 | NP_001009955.1 | 10, 11 | 1066 | ||
NM_018070.4 | NP_060540.2 | 10, 11 | 1087 | |||
NM_145716.3 | NP_663768.1 | 11, 12 | 1147 | |||
XM_006710545.3 | XP_006710608.1 | 9, 10 | 646 | |||
XM_017000897.1 | XP_016856386.1 | 11, 12 | 816 | |||
XM_017000898.1 | XP_016856387.1 | 8, 9 | 1043 | |||
XM_017000899.1 | XP_016856388.1 | 10, 11 | 747 | |||
GenBank mRNA | AF500116.1 | - | 10, 11 | 915 | ||
AK001217.1 | - | 7, 8 | 495 | |||
AK093975.1 | - | 7, 8 | 807 | |||
AK289474.1 | - | 10, 11 | 688 | |||
BC003605.1 | - | 10, 11 | 726 | 3546126 | ||
BC004335.1 | - | 4, 5 | 244 | |||
BC066365.1 | - | 11, 12 | 891 | 6169677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
