siRNA Details
Gene Symbol: SSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009955.3 | NP_001009955.1 | 17 | 1659 | ||
NM_018070.4 | NP_060540.2 | 17 | 1680 | |||
NM_145716.3 | NP_663768.1 | 18 | 1740 | |||
XM_006710545.3 | XP_006710608.1 | 16 | 1239 | |||
XM_017000897.1 | XP_016856386.1 | 18 | 1409 | |||
XM_017000898.1 | XP_016856387.1 | 15 | 1636 | |||
XM_017000899.1 | XP_016856388.1 | 17 | 1340 | |||
GenBank mRNA | AF086085.1 | - | 4 | 378 | ||
AF500116.1 | - | 17 | 1508 | |||
AK001217.1 | - | 14 | 1088 | |||
AK093975.1 | - | 14 | 1400 | |||
AK289474.1 | - | 17 | 1281 | |||
BC003605.1 | - | 17 | 1319 | 3546126 | ||
BC004335.1 | - | 11 | 837 | |||
BC066365.1 | - | 18 | 1484 | 6169677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
