siRNA Details
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173619.3 | NP_775890.2 | 12 | 1702 | ||
XM_006721179.1 | XP_006721242.1 | 12 | 1798 | |||
XM_011523034.1 | XP_011521336.1 | 13 | 2044 | |||
XM_011523036.2 | XP_011521338.1 | 11 | 1628 | |||
GenBank mRNA | BC039068.1 | - | 10 | 2756 | 5188643 |
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC078143.1 | - | 13 | 1716 | 5761001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
