siRNA Details
Gene Symbol: SMG1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135305.1 | 11 | 1449 | |||
NR_135317.1 | 9 | 1644 | ||||
GenBank mRNA | AB055226.1 | - | 11 | 1860 | ||
AK097897.1 | - | 9 | 1155 |
Gene Symbol: SMG1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027154.1 | 10 | 1667 |
Gene Symbol: SMG1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027155.2 | 10 | 1687 | |||
GenBank mRNA | BC055086.1 | - | 1 | 15 |
Gene Symbol: SMG1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002453.5 | 10 | 1585 | |||
GenBank mRNA | BC063703.1 | - | 10 | 1585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
