siRNA Details
Gene Symbol: SMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015092.4 | NP_055907.3 | 11 | 1791 | ||
XM_005255182.3 | XP_005255239.1 | 11 | 1804 | |||
XM_005255183.3 | XP_005255240.1 | 12 | 1862 | |||
XM_005255184.3 | XP_005255241.1 | 10 | 1648 | |||
XM_011545769.2 | XP_011544071.1 | 11 | 1804 | |||
XM_017023066.1 | XP_016878555.1 | 11 | 1614 | |||
GenBank mRNA | AB061371.1 | - | 12 | 1743 | ||
AF395444.1 | - | 9 | 1098 |
Gene Symbol: BOLA2-SMG1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320622.1 | NP_001307551.1 | 3 | 969 | ||
NM_001320623.1 | NP_001307552.1 | 3 | 903 | |||
NM_001320624.1 | NP_001307553.1 | 2 | 909 | |||
NM_001320625.1 | NP_001307554.1 | 2 | 805 | |||
NM_001320627.1 | NP_001307556.1 | 3 | 1007 | |||
NM_001320628.1 | NP_001307557.1 | 2 | 325 | |||
GenBank mRNA | AK027255.1 | - | 4 | 2471 | ||
AK295153.1 | - | 3 | 311 | |||
BC062756.1 | - | 3 | 388 | |||
BG030350.1 | - | 260 | ||||
BM451872.1 | - | 3 | 347 | |||
BM696373.1 | - | 2 | 310 | |||
DA438012.1 | - | 3 | 295 |
Gene Symbol: LOC107984138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023965.1 | XP_016879454.1 | 4 | 479 | ||
XM_017023966.1 | XP_016879455.1 | 4 | 479 | |||
XM_017023967.1 | XP_016879456.1 | 4 | 479 |
Gene Symbol: SMG1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027154.1 | 11 | 1817 |
Gene Symbol: SMG1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135305.1 | 12 | 1599 | |||
NR_135317.1 | 10 | 1794 | ||||
GenBank mRNA | AB055226.1 | - | 12 | 2010 | ||
AK097897.1 | - | 10 | 1305 | |||
BX640895.1 | - | 1 | 77 |
Gene Symbol: SMG1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027155.2 | 11 | 1837 | |||
GenBank mRNA | BC055086.1 | - | 2 | 165 |
Gene Symbol: SMG1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002453.5 | 11 | 1735 | |||
GenBank mRNA | BC063703.1 | - | 11 | 1735 |
Gene Symbol: SMG1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135312.1 | 1 | 135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
