siRNA Details
Gene Symbol: EIF1AY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278612.1 | NP_001265541.1 | 1, 2 | 177 | ||
NM_004681.3 | NP_004672.2 | 1, 2 | 177 | |||
GenBank mRNA | AF000987.1 | - | 1, 2 | 148 | ||
BC005248.1 | - | 1, 2 | 59 | 3933718 | ||
BF694967.1 | - | 164 | ||||
BG499107.1 | - | 1, 2 | 163 | |||
BP331805.1 | - | 1, 2 | 177 | |||
BT007201.1 | - | 1, 2 | 16 | |||
DQ786267.1 | - | 1, 2 | 100 |
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001412.3 | NP_001403.1 | 1, 2 | 224 | ||
GenBank mRNA | AK312320.1 | - | 1 | 211 | ||
BC000793.1 | - | 1, 2 | 185 | 3451631 | ||
BC067851.1 | - | 1, 2 | 211 | 5288453 | ||
BC110421.1 | - | 1, 2 | 189 | |||
BT007064.1 | - | 1 | 16 | |||
L18960.1 | - | 1, 2 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
