siRNA Details
Gene Symbol: EIF1AY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278612.1 | NP_001265541.1 | 2 | 223 | ||
NM_004681.3 | NP_004672.2 | 2 | 223 | |||
GenBank mRNA | AF000987.1 | - | 2 | 194 | ||
BC005248.1 | - | 2 | 105 | 3933718 | ||
BF694967.1 | - | 210 | ||||
BP331805.1 | - | 2 | 223 | |||
BT007201.1 | - | 2 | 62 | |||
DQ786267.1 | - | 2 | 146 |
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001412.3 | NP_001403.1 | 2 | 270 | ||
GenBank mRNA | AK312320.1 | - | 1 | 257 | ||
BC000793.1 | - | 2 | 231 | 3451631 | ||
BC067851.1 | - | 2 | 257 | 5288453 | ||
BC110421.1 | - | 2 | 235 | |||
BT007064.1 | - | 1 | 62 | |||
L18960.1 | - | 2 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
