siRNA Details
Gene Symbol: TMSB15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021992.2 | NP_068832.1 | 2 | 149 | ||
GenBank mRNA | AF086495.1 | - | 2 | 92 | ||
BC000183.1 | - | 2 | 97 | 3350388 | ||
BG829126.1 | - | 2 | 99 | |||
BU599402.1 | - | 2 | 162 | |||
D82345.1 | - | 2 | 123 |
Gene Symbol: LOC100101478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028039.1 | - | 2 | 135 |
Gene Symbol: TMSB15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194324.2 | NP_919305.2 | 2 | 140 | ||
GenBank mRNA | AK291479.1 | - | 2 | 116 | ||
AK311771.1 | - | 2 | 123 | |||
BC093093.1 | - | 2 | 140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
