siRNA Details
Gene Symbol: TUBB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177987.2 | NP_817124.1 | 4 | 1259 | ||
XM_011519459.2 | XP_011517761.1 | 4 | 1299 | |||
XM_011519460.2 | XP_011517762.1 | 3 | 945 | |||
XM_017016192.1 | XP_016871681.1 | 2 | 1196 | |||
XM_017016193.1 | XP_016871682.1 | 2 | 1016 | |||
GenBank mRNA | BC101270.2 | - | 4 | 1252 | ||
BC101271.1 | - | 4 | 1252 |
Gene Symbol: TUBB8P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014971.1 | - | 3 | 1876 | ||
BC139719.1 | - | 4 | 1450 |
Gene Symbol: TUBBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027156.1 | 5 | 2415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
