siRNA Details
Gene Symbol: SHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301168.1 | NP_001288097.1 | 3 | 780 | ||
NM_001301169.1 | NP_001288098.1 | 3 | 780 | |||
NM_001301170.1 | NP_001288099.1 | 3 | 780 | |||
NM_001301171.1 | NP_001288100.1 | 3 | 367 | |||
NM_138356.2 | NP_612365.2 | 4 | 1064 | |||
XM_006720742.3 | XP_006720805.2 | 3 | 779 | |||
XM_011522174.2 | XP_011520476.1 | 3 | 779 | |||
XM_011522175.2 | XP_011520477.1 | 3 | 779 | |||
XM_011522176.2 | XP_011520478.1 | 3 | 779 | |||
XM_011522177.2 | XP_011520479.1 | 3 | 779 | |||
XM_011522178.2 | XP_011520480.1 | 3 | 779 | |||
XM_017022710.1 | XP_016878199.1 | 3 | 780 | |||
XM_017022711.1 | XP_016878200.1 | 3 | 890 | |||
GenBank mRNA | AK094391.1 | - | 3 | 508 | ||
AK096267.1 | - | 4 | 1064 | |||
AK301625.1 | - | 3 | 367 | |||
AK303749.1 | - | 4 | 555 | |||
BC007586.1 | - | 2 | 142 | |||
BC101601.1 | - | 2 | 220 | 8069107 | ||
BC101603.1 | - | 2 | 219 | 8069109 | ||
BC143601.1 | - | 2 | 220 | |||
BC171730.1 | - | 2 | 220 | |||
BX446025.2 | - | 3 | 465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
