siRNA Details
Gene Symbol: TCAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130026.2 | NP_001123498.2 | 8 | 2249 | ||
XM_006715927.3 | XP_006715990.1 | 7 | 3481 | |||
XM_011516059.2 | XP_011514361.1 | 7 | 2069 | |||
GenBank mRNA | AK090395.1 | - | 6 | 1712 | ||
AK296318.1 | - | 8 | 2225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
