siRNA Details
Gene Symbol: RPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031.4 | NP_001022.1 | 1 | 45 | ||
GenBank mRNA | AK293636.1 | - | 1 | 45 | ||
AK311925.1 | - | 1 | 45 | |||
AU126783.1 | - | 1 | 45 | |||
BC000354.1 | - | 1 | 31 | 2964364 | ||
BC018810.1 | - | 1 | 31 | |||
BC021239.2 | - | 1 | 24 | 4309752 | ||
BC070217.1 | - | 1 | 45 | 4248660 | ||
BC070218.1 | - | 1 | 42 | 6649143 | ||
CR457055.1 | - | 1 | 14 | |||
DQ586798.1 | - | 1 | 13 | |||
L05091.1 | - | 1 | 29 | |||
U58682.1 | - | 1 | 26 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
