siRNA Details
Gene Symbol: BCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004327.3 | NP_004318.3 | 20, 21 | 4054 | ||
NM_021574.2 | NP_067585.2 | 19, 20 | 3922 | |||
GenBank mRNA | AB209991.1 | - | 20, 21 | 3803 | ||
AK122842.1 | - | 3, 4 | 1318 | |||
AK303241.1 | - | 20, 21 | 2488 | |||
BC066122.1 | - | 20, 21 | 3885 | |||
X02596.1 | - | 20, 21 | 3946 | |||
Y00661.1 | - | 19, 20 | 3922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
