siRNA Details
Gene Symbol: RPL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000992.2 | NP_000983.1 | 4 | 242 | ||
GenBank mRNA | AK289538.1 | - | 4 | 195 | ||
AK311884.1 | - | 4 | 195 | |||
BC008926.2 | - | 4 | 242 | 3835176 | ||
BC018943.2 | - | 1 | 20 | |||
BC070190.1 | - | 1 | 193 | 6452382 | ||
BC070481.1 | - | 4 | 181 | 6025704 | ||
BC071663.1 | - | 1 | 167 | 5220693 | ||
BC071909.1 | - | 4 | 180 | 6729601 | ||
U10248.1 | - | 4 | 177 | |||
U49083.1 | - | 4 | 175 | |||
Z49148.1 | - | 4 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
