siRNA Details
Gene Symbol: TMEM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322248.1 | NP_001309177.1 | 3 | 2591 | ||
NM_020141.3 | NP_064526.1 | 3 | 2490 | |||
NR_136242.1 | 4 | 2619 | ||||
NR_136243.1 | 2 | 2398 | ||||
GenBank mRNA | BC008739.2 | - | 3 | 2490 | 3346340 | |
BC065186.1 | - | 3 | 2490 | 4642864 | ||
BC066918.1 | - | 3 | 2519 | 4826492 | ||
BC067116.1 | - | 3 | 2476 | 5532892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
