siRNA Details
Gene Symbol: LSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013253.1 | NP_001013271.1 | 9 | 1137 | ||
NM_001013254.1 | NP_001013272.1 | 9 | 1367 | |||
NM_001013255.1 | NP_001013273.1 | 9 | 873 | |||
NM_001242932.1 | NP_001229861.1 | 10 | 1368 | |||
NM_001289005.1 | NP_001275934.1 | 9 | 1233 | |||
NM_002339.2 | NP_002330.1 | 9 | 1051 | |||
GenBank mRNA | AK056576.1 | - | 9 | 1992 | ||
AK092071.1 | - | 10 | 1368 | |||
AK093859.1 | - | 8 | 1374 | |||
AK222733.1 | - | 9 | 947 | |||
AK290279.1 | - | 9 | 889 | |||
AK292033.1 | - | 9 | 1235 | |||
BC001785.1 | - | 9 | 943 | 3542307 | ||
CR541728.1 | - | 9 | 876 | |||
CR541754.1 | - | 9 | 876 | |||
S67783.1 | - | 9 | 873 | |||
X55188.1 | - | 9 | 923 |
Gene Symbol: LOC107985911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005465.1 | XP_016860954.1 | 3 | 571 |
Gene Symbol: LOC645166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027354.2 | 3 | 332 | |||
NR_027355.2 | 3 | 232 | ||||
GenBank mRNA | AK311729.1 | - | 3 | 332 | ||
BE814857.1 | - | 3 | 224 | |||
BQ438821.1 | - | 3 | 233 | |||
CR746216.1 | - | 3 | 259 |
Gene Symbol: LOC654342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027238.1 | 3 | 200 | |||
GenBank mRNA | BF031534.1 | - | 3 | 200 | ||
BM826054.1 | - | 2 | 59 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
