siRNA Details
Gene Symbol: LSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013253.1 | NP_001013271.1 | 8 | 1027 | ||
NM_001013254.1 | NP_001013272.1 | 8 | 1257 | |||
NM_001013255.1 | NP_001013273.1 | 8 | 763 | |||
NM_001242932.1 | NP_001229861.1 | 9 | 1258 | |||
NM_001289005.1 | NP_001275934.1 | 8 | 1123 | |||
NM_002339.2 | NP_002330.1 | 8 | 941 | |||
GenBank mRNA | AK056576.1 | - | 8 | 1882 | ||
AK092071.1 | - | 9 | 1258 | |||
AK093859.1 | - | 7 | 1264 | |||
AK222733.1 | - | 8 | 837 | |||
AK290279.1 | - | 8 | 779 | |||
AK292033.1 | - | 8 | 1125 | |||
BC001785.1 | - | 8 | 833 | 3542307 | ||
BQ893037.1 | - | 8 | 802 | |||
CR541728.1 | - | 8 | 766 | |||
CR541754.1 | - | 8 | 766 | |||
S67783.1 | - | 8 | 763 | |||
X55188.1 | - | 8 | 813 |
Gene Symbol: LOC107985911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005465.1 | XP_016860954.1 | 2 | 461 |
Gene Symbol: LOC645166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027354.2 | 2 | 222 | |||
NR_027355.2 | 2 | 122 | ||||
NR_027356.2 | 2 | 122 | ||||
GenBank mRNA | AK311729.1 | - | 2 | 222 | ||
AW954772.1 | - | 2 | 111 | |||
BG716766.1 | - | 2 | 114 | |||
BQ438821.1 | - | 2 | 123 | |||
DA808118.1 | - | 2 | 124 | |||
DB496177.1 | - | 2 | 109 |
Gene Symbol: LOC654342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027238.1 | 2 | 90 | |||
GenBank mRNA | BC030764.1 | - | 2 | 114 | ||
BF031534.1 | - | 2 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
