siRNA Details
Gene Symbol: MT1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005951.2 | NP_005942.1 | 2 | 138 | ||
GenBank mRNA | BC059780.1 | - | 2 | 118 | 5227592 | |
BC146930.1 | - | 2 | 89 | 9056858 | ||
BC146934.1 | - | 2 | 89 | 9056862 | ||
S68948.1 | - | 2 | 67 | |||
X64177.1 | - | 2 | 90 |
Gene Symbol: MT1HL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276687.1 | NP_001263616.1 | 1 | 113 | ||
GenBank mRNA | AF333388.1 | - | 1 | 113 | ||
BC133030.1 | - | 1 | 107 | |||
BC133032.1 | - | 1 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
