siRNA Details
Gene Symbol: CCDC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258304.1 | NP_001245233.1 | 3 | 630 | ||
NM_001258305.1 | NP_001245234.1 | 3 | 504 | |||
NM_138770.2 | NP_620125.1 | 3 | 504 | |||
XM_005263836.4 | XP_005263893.1 | 1 | 787 | |||
XM_005263837.1 | XP_005263894.1 | 3 | 422 | |||
XM_006712828.3 | XP_006712891.1 | 3 | 603 | |||
XM_017005234.1 | XP_016860723.1 | 3 | 604 | |||
XM_017005235.1 | XP_016860724.1 | 3 | 663 | |||
XM_017005237.1 | XP_016860726.1 | 3 | 619 | |||
XM_017005238.1 | XP_016860727.1 | 3 | 628 | |||
XM_017005240.1 | XP_016860729.1 | 3 | 663 | |||
XM_017005241.1 | XP_016860730.1 | 4 | 400 | |||
XM_017005245.1 | XP_016860734.1 | 3 | 422 | |||
GenBank mRNA | AB299086.1 | - | 3 | 545 | ||
AB299087.1 | - | 3 | 419 | |||
AK097664.1 | - | 1 | 987 | |||
BC016861.1 | - | 3 | 419 | 3914851 | ||
CV023784.1 | - | 3 | 367 |
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207310.2 | NP_997193.1 | 3 | 505 | ||
XM_005263840.2 | XP_005263897.1 | 3 | 530 | |||
XM_006712833.2 | XP_006712896.1 | 3 | 530 | |||
XM_006712834.3 | XP_006712897.1 | 3 | 470 | |||
XM_011512142.2 | XP_011510444.1 | 1 | 773 | |||
XM_011512147.1 | XP_011510449.1 | 3 | 530 | |||
GenBank mRNA | AK057609.1 | - | 3 | 429 | ||
AK297246.1 | - | 2 | 1417 | |||
AL133619.1 | - | 1 | 719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
