siRNA Details
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258307.1 | NP_001245236.1 | 2 | 407 | ||
NM_207310.2 | NP_997193.1 | 2 | 407 | |||
XM_005263840.2 | XP_005263897.1 | 2 | 432 | |||
XM_005263842.2 | XP_005263899.1 | 2 | 432 | |||
XM_006712833.2 | XP_006712896.1 | 2 | 432 | |||
XM_006712834.3 | XP_006712897.1 | 2 | 372 | |||
XM_006712835.2 | XP_006712898.1 | 2 | 432 | |||
XM_006712837.2 | XP_006712900.1 | 2 | 372 | |||
XM_011512147.1 | XP_011510449.1 | 2 | 432 | |||
GenBank mRNA | AK057609.1 | - | 2 | 331 | ||
AK297246.1 | - | 2 | 432 | |||
AK302804.1 | - | 2 | 356 | |||
BC067771.1 | - | 2 | 352 | 4824929 |
Gene Symbol: CCDC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258304.1 | NP_001245233.1 | 2 | 532 | ||
NM_001258305.1 | NP_001245234.1 | 2 | 406 | |||
NM_001258306.1 | NP_001245235.1 | 2 | 406 | |||
NM_138770.2 | NP_620125.1 | 2 | 406 | |||
XM_005263835.3 | XP_005263892.1 | 2 | 504 | |||
XM_005263837.1 | XP_005263894.1 | 2 | 352 | |||
XM_006712828.3 | XP_006712891.1 | 2 | 505 | |||
XM_017005234.1 | XP_016860723.1 | 2 | 506 | |||
XM_017005235.1 | XP_016860724.1 | 2 | 565 | |||
XM_017005236.1 | XP_016860725.1 | 2 | 505 | |||
XM_017005237.1 | XP_016860726.1 | 2 | 521 | |||
XM_017005238.1 | XP_016860727.1 | 2 | 530 | |||
XM_017005239.1 | XP_016860728.1 | 2 | 565 | |||
XM_017005240.1 | XP_016860729.1 | 2 | 565 | |||
XM_017005241.1 | XP_016860730.1 | 3 | 302 | |||
XM_017005242.1 | XP_016860731.1 | 2 | 513 | |||
XM_017005243.1 | XP_016860732.1 | 2 | 524 | |||
XM_017005244.1 | XP_016860733.1 | 3 | 296 | |||
XM_017005245.1 | XP_016860734.1 | 2 | 352 | |||
XM_017005246.1 | XP_016860735.1 | 2 | 406 | |||
GenBank mRNA | AB299086.1 | - | 2 | 447 | ||
AB299087.1 | - | 2 | 321 | |||
AK309113.1 | - | 2 | 565 | |||
BC016861.1 | - | 2 | 321 | 3914851 | ||
BC063387.1 | - | 2 | 343 | 5494806 | ||
CV023784.1 | - | 2 | 269 | |||
DA375407.1 | - | 2 | 406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
